Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aman-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374889.1 | NP_001361919.1 | 17 - 18 | 2532 | 70 | |
NM_001374890.2 | NP_001362173.1 | 17 - 18 | 2536 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |