Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059179.3 | NP_491580.1 | 2 - 3 | 481 | 80 | |
NM_059180.2 | NP_491581.1 | 2 - 3 | 360 | 80 | ||
NM_170865.5 | NP_740852.1 | 3 - 4 | 379 | 80 | ||
NM_182025.7 | NP_871825.1 | 2 - 3 | 266 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |