Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026479.4 | NP_001021650.1 | 23 - 24 | 5660 | 91 | |
NM_001381215.1 | NP_001366962.1 | 3 - 4 | 825 | 91 | ||
GenBank mRNA | AJ307058.1 | - | 23 - 24 | 5660 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |