Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047328.5 | NP_001040793.1 | 7 - 8 | 2347 | 101 | |
NM_001047329.4 | NP_001040794.1 | 7 - 8 | 2191 | 101 | ||
NM_001313645.3 | NP_001300574.1 | 4 - 5 | 1864 | 101 | ||
NM_001381394.1 | NP_001367817.1 | 8 - 9 | 2396 | 101 | ||
NM_001381395.2 | NP_001367816.1 | 8 - 9 | 2400 | 101 | ||
NM_170979.6 | NP_740976.1 | 5 - 6 | 2302 | 101 | ||
GenBank mRNA | EU180219.1 | - | 8 - 9 | 2380 | 101 |
Gene Symbol: coa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383820.1 | 10 - 11 | 2985 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |