Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgt-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381440.1 | NP_001367789.1 | 3 - 4 | 722 | 117 | |
NM_062780.6 | NP_495181.2 | 5 - 6 | 893 | 117 | ||
NM_062781.5 | NP_495182.2 | 4 - 5 | 817 | 117 | ||
NR_151564.1 | 4 - 5 | 786 | 117 | |||
GenBank mRNA | DQ178632.1 | - | 722 | 117 | ||
DQ178633.1 | - | 893 | 117 | |||
DQ178634.1 | - | 833 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |