Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381467.2 | NP_001367422.1 | 5 - 6 | 860 | 107 | |
NM_062999.9 | NP_495400.2 | 5 - 6 | 847 | 107 | ||
NM_171014.6 | NP_741015.1 | 5 - 6 | 844 | 107 | ||
NM_171015.6 | NP_741016.1 | 5 - 6 | 831 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |