Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sax-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171117.5 | NP_741131.2 | 28 - 29 | 7871 | 64 | |
NM_171118.6 | NP_741132.2 | 3 - 4 | 1355 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |