Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171123.6 | NP_741140.1 | 6 - 7 | 1164 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |