Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027422.4 | NP_001022593.1 | 3 - 4 | 437 | 100 | |
NM_001027423.6 | NP_001022594.1 | 3 - 4 | 449 | 100 | ||
NM_001027425.5 | NP_001022596.1 | 3 - 4 | 394 | 100 | ||
NM_001027426.4 | NP_001022597.1 | 3 - 4 | 451 | 100 | ||
NM_001379757.2 | NP_001368076.1 | 5 - 6 | 573 | 100 | ||
GenBank mRNA | AF272738.1 | - | 3 - 4 | 394 | 100 | |
AY053364.1 | - | 3 - 4 | 394 | 100 | ||
AY053365.1 | - | 3 - 4 | 394 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |