Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171180.5 | NP_741210.3 | 5 - 6 | 1063 | 67 | |
NM_171880.6 | NP_741211.3 | 5 - 6 | 1065 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |