Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382930.1 | 7 - 8 | 2027 | 74 | ||
NM_171203.6 | NP_741240.1 | 7 - 8 | 2025 | 74 | ||
NM_171205.6 | NP_741242.1 | 4 - 5 | 1332 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |