Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hipr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066524.6 | NP_498925.1 | 2 - 3 | 532 | 103 | |
NM_171215.7 | NP_741253.1 | 2 - 3 | 533 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |