Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027810.5 | NP_001022981.1 | 15 - 16 | 2627 | 76 | |
NM_001129275.4 | NP_001122747.1 | 5 - 6 | 730 | 76 | ||
NM_001129276.3 | NP_001122748.1 | 16 - 17 | 2711 | 76 | ||
NM_001268079.4 | NP_001255008.1 | 16 - 17 | 2726 | 76 | ||
NM_001268080.3 | NP_001255009.1 | 11 - 12 | 1816 | 76 | ||
NM_001306679.3 | NP_001293608.1 | 9 - 10 | 1612 | 76 | ||
NM_001306687.3 | NP_001293616.1 | 10 - 11 | 1630 | 76 | ||
NM_001306688.3 | NP_001293617.1 | 6 - 7 | 814 | 76 | ||
NM_001382965.1 | 18 - 19 | 3032 | 76 | |||
NM_171221.7 | NP_741263.3 | 17 - 18 | 2718 | 76 | ||
GenBank mRNA | AF424978.1 | - | 15 - 16 | 2563 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |