Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171226.5 | NP_741268.1 | 2 - 3 | 219 | 83 | |
NM_171892.7 | NP_741269.1 | 2 - 3 | 200 | 83 | ||
NM_171893.6 | NP_741270.1 | 2 - 3 | 207 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |