Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171226.5 | NP_741268.1 | 4 - 5 | 1099 | 62 | |
NM_171892.7 | NP_741269.1 | 4 - 5 | 1080 | 62 | ||
NM_171893.6 | NP_741270.1 | 4 - 5 | 1087 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |