Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elo-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268181.3 | NP_001255110.1 | 4 - 5 | 632 | 71 | |
NM_001268182.3 | NP_001255111.1 | 1 - 2 | 44 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |