Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: seu-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380269.2 | NP_001368401.1 | 3 - 4 | 1450 | 138 | |
NM_001380270.1 | NP_001368061.1 | 4 - 5 | 1102 | 138 | ||
NM_068564.7 | NP_500965.1 | 7 - 8 | 1642 | 138 | ||
NM_068565.7 | NP_500966.1 | 6 - 7 | 1507 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |