Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306912.2 | NP_001293841.1 | 9 - 10 | 1807 | 109 | |
NM_001306913.3 | NP_001293842.1 | 10 - 11 | 1873 | 109 | ||
NM_001306914.2 | NP_001293843.1 | 9 - 10 | 1771 | 109 | ||
NM_001380288.1 | NP_001368430.1 | 8 - 9 | 1572 | 109 | ||
NM_001383109.1 | 10 - 11 | 1830 | 109 | |||
NM_171367.8 | NP_741430.3 | 9 - 10 | 1704 | 109 | ||
GenBank mRNA | AY455928.1 | - | 9 - 10 | 1766 | 109 | |
AY493413.1 | - | 9 - 10 | 1758 | 109 | ||
AY493414.1 | - | 8 - 9 | 1587 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |