Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027908.6 | NP_001023079.1 | 4 - 5 | 506 | 128 | |
NM_171379.6 | NP_741442.1 | 4 - 5 | 506 | 128 | ||
GenBank mRNA | AF023602.1 | - | 5 - 6 | 539 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |