Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171383.5 | NP_741446.1 | 3 - 4 | 290 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |