Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361754.3 | NP_001348668.1 | 3 - 4 | 735 | 68 | |
NM_059465.5 | NP_491866.2 | 3 - 4 | 736 | 68 | ||
GenBank mRNA | AY887907.1 | - | 3 - 4 | 752 | 68 | |
AY954523.1 | - | 3 - 4 | 752 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |