Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059517.4 | NP_491918.3 | 8 - 9 | 1016 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |