Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383275.1 | 3 - 4 | 270 | 89 | ||
GenBank mRNA | AF295927.1 | - | 2 - 3 | 168 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |