Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025858.4 | NP_001021029.1 | 3 - 4 | 794 | 97 | |
NM_001306351.3 | NP_001293280.1 | 1 - 2 | 132 | 97 | ||
NM_059539.6 | NP_491940.2 | 3 - 4 | 553 | 97 | ||
GenBank mRNA | AF170425.1 | - | 3 - 4 | 563 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |