Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026410.4 | NP_001021581.1 | 4 - 5 | 1095 | 138 | |
NR_002347.1 | 5 - 6 | 1034 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |