Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpg-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059646.7 | NP_492047.1 | 3 - 4 | 621 | 69 | |
GenBank mRNA | DQ340625.1 | - | 2 - 3 | 425 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |