Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059671.9 | NP_492072.2 | 4 - 5 | 566 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |