Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059671.9 | NP_492072.2 | 7 - 8 | 1076 | 72 | |
NR_132562.1 | 1 - 2 | 98 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |