Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059687.7 | NP_492088.3 | 9 - 10 | 1222 | 152 | |
GenBank mRNA | AY387697.1 | - | 9 - 10 | 1187 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |