Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 26 - 27 | 5112 | 70 | |
NM_001026702.3 | NP_001021873.1 | 28 - 29 | 6138 | 70 | ||
NM_001026703.4 | NP_001021874.1 | 27 - 28 | 5121 | 70 | ||
NM_001263573.3 | NP_001250502.1 | 27 - 28 | 5130 | 70 | ||
NM_001306224.3 | NP_001293153.1 | 15 - 16 | 4210 | 70 | ||
NM_001330927.3 | NP_001317864.1 | 26 - 27 | 5121 | 70 | ||
NM_001330928.3 | NP_001317865.1 | 26 - 27 | 5076 | 70 | ||
NM_001330929.3 | NP_001317866.1 | 26 - 27 | 5067 | 70 | ||
NM_001330930.3 | NP_001317863.1 | 18 - 19 | 4089 | 70 | ||
NM_001330931.3 | NP_001317867.1 | 14 - 15 | 4068 | 70 | ||
NM_001380703.1 | NP_001367037.1 | 19 - 20 | 4098 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |