Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nmy-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059785.9 | NP_492186.3 | 12 - 13 | 4046 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |