Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136304.4 | NP_001129776.1 | 3 - 4 | 864 | 166 | |
NM_001380825.1 | NP_001366789.1 | 3 - 4 | 387 | 166 | ||
NM_001380826.1 | NP_001367194.1 | 2 - 3 | 463 | 166 | ||
GenBank mRNA | AY887903.1 | - | 3 - 4 | 409 | 166 | |
DQ178237.1 | - | 2 - 3 | 545 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |