Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136304.4 | NP_001129776.1 | 5 - 6 | 1164 | 114 | |
NM_001380825.1 | NP_001366789.1 | 5 - 6 | 687 | 114 | ||
NM_001380826.1 | NP_001367194.1 | 5 - 6 | 925 | 114 | ||
GenBank mRNA | AY887903.1 | - | 5 - 6 | 709 | 114 | |
DQ178237.1 | - | 4 - 5 | 845 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |