Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sin-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128970.4 | NP_001122442.1 | 14 - 15 | 3299 | 108 | |
NM_059883.8 | NP_492284.2 | 14 - 15 | 3294 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |