Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059911.2 | NP_492312.1 | 2 - 3 | 300 | 93 | |
GenBank mRNA | AY008137.1 | - | 1 - 2 | 169 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |