Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373211.3 | NP_001359628.1 | 2 - 3 | 326 | 132 | |
NM_059972.6 | NP_492373.1 | 3 - 4 | 410 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |