Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373211.3 | NP_001359628.1 | 7 - 8 | 1238 | 130 | |
NM_059972.6 | NP_492373.1 | 8 - 9 | 1322 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |