Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sup-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059976.7 | NP_492377.1 | 6 - 7 | 638 | 115 | |
GenBank mRNA | AF024614.1 | - | 6 - 7 | 629 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |