Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sup-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059976.7 | NP_492377.1 | 9 - 10 | 1222 | 100 | |
GenBank mRNA | AF024614.1 | - | 9 - 10 | 1213 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |