Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rad-54.L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060037.5 | NP_492438.1 | 14 - 15 | 2189 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |