Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026506.5 | NP_001021677.3 | 9 - 10 | 1298 | 83 | |
NM_001026507.5 | NP_001021678.3 | 9 - 10 | 1550 | 83 | ||
NM_001026508.6 | NP_001021679.3 | 8 - 9 | 1098 | 83 | ||
NM_060041.6 | NP_492442.3 | 9 - 10 | 1284 | 83 | ||
NM_060042.5 | NP_492443.3 | 9 - 10 | 1270 | 83 | ||
NM_060043.5 | NP_492444.4 | 9 - 10 | 1449 | 83 | ||
NR_000051.1 | 10 - 11 | 1468 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |