Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313465.3 | NP_001300394.1 | 3 | 427 | 112 | |
NM_001313466.5 | NP_001300395.1 | 4 | 517 | 112 | ||
NM_001377616.3 | NP_001364651.1 | 1 - 2 | 106 | 112 | ||
GenBank mRNA | EF443133.1 | - | 1 - 2 | 106 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |