Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 10 - 11 | 1627 | 66 | |
NM_001026218.5 | NP_001021389.1 | 10 - 11 | 1633 | 66 | ||
NM_001026219.5 | NP_001021390.1 | 10 - 11 | 1578 | 66 | ||
NM_001128984.4 | NP_001122456.2 | 12 - 13 | 1794 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |