Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ile-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060147.6 | NP_492548.1 | 5 - 6 | 1235 | 121 | |
GenBank mRNA | DQ384615.1 | - | 5 - 6 | 1233 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |