Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264316.2 | NP_001251245.1 | 3 - 4 | 585 | 97 | |
NM_001264317.3 | NP_001251246.1 | 2 - 3 | 276 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |