Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060234.7 | NP_492635.1 | 1 - 2 | 533 | 78 | |
GenBank mRNA | AF013953.1 | - | 1 - 2 | 525 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |