Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aha-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264398.2 | NP_001251327.1 | 5 - 6 | 830 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |