Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060307.4 | NP_492708.1 | 1 - 2 | 152 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |