Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: duo-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306534.2 | NP_001293463.1 | 10 - 11 | 2551 | 69 | |
NM_001306535.3 | NP_001293464.1 | 7 - 8 | 2086 | 69 | ||
NM_001306536.3 | NP_001293465.1 | 5 - 6 | 1603 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |