Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: duo-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306534.2 | NP_001293463.1 | 4 - 5 | 693 | 124 | |
NM_001306535.3 | NP_001293464.1 | 1 - 2 | 228 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |